1-Iodoperfluorbutane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane
A16671-22
A16671-06
A16671-14
CAAAAA16671-22EA
267.02
CAD
CAAAAA16671-22
CAAAA16671-06
CAAAA16671-14
1-Iodoperfluorbutane 97%
1-Iodoperfluorbutane
Purity: 97%
CAS No.: 423-39-2
Molecular Formula: CF3(CF2)3I
Formula Weight: 345.93
Refractive Index: 1.3290
Odor: Pungent
Boiling Point: 67°
Beilstein Reference: 1777546
UN No.: 3082
MDL No.: MFCD00001062
Formula:
C₄F₉I Boiling Pt: 67 °C Melting Pt: –89 °C…–87 °C Density: 2.010 Storage Temperature: Ambient |
MDL Number:
MFCD00001062 CAS Number: 423-39-2 EINECS: 207-025-8 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...