3-Chloro-4-fluorobenzotrifluoride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Chloro-2-fluoro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluorobenzotrifluoride, 2-Chloro-1-fluoro-4-(trifluoromethyl)benzene
A16670-06
A16670-14
CAAAA16670-06EA
140.15
CAD
CAAAA16670-06
CAAAA16670-14
3-Chloro-4-fluorobenzotrifluoride 98%
3-Chloro-4-fluorobenzotrifluoride
Purity: 98%
CAS No.: 78068-85-6
Molecular Formula: C7H3ClF4
Formula Weight: 198.55
Flash Point: 42°(108°F)
Refractive Index: 1.4355
Boiling Point: 137°
Beilstein Reference: 4309386
UN No.: 1993
MDL No.: MFCD00042207
Formula:
C₇H₃ClF₄ MW: 198.55 g/mol Boiling Pt: 137 °C Density: 1.490 Flash Pt: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042207 CAS Number: 78068-85-6 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...