β-Alanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
A16665-0B
A16665-0I
A16665-30
CAAAA16665-0BEA
132.25
CAD
CAAAA16665-0B
CAAAA16665-0I
CAAAA16665-30
β-Alanine 98%
β-Alanine
Purity: 98%
CAS No.: 107-95-9
Molecular Formula: C3H7NO2
Formula Weight: 89.09
Physical Form: Cryst powder
Density: 1.437
Appearance: White
Odor: Odorless
Melting Point: ca 205° dec.
Beilstein Reference: 906793
Merck Reference: 14,205
MDL No.: MFCD00008200
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 201 Melting Pt: 94 (dec.) Density: 1.437 Storage Temperature: Ambient |
MDL Number:
MFCD00008200 CAS Number: 107-95-9 EINECS: 203-536-5 Merck Index: 14,205 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...