Oleic acid tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
A16663-0J
A16663-AU
A16663-0J
CAAAA16663-0JEA
222.78
CAD
CAAAA16663-0J
CAAAA16663-AU
AAA16663-0J
Oleic acid tech. 90%, Technical Grade
Oleic acid
Purity: 90%
CAS No.: 112-80-1
Molecular Formula: C18H34O2
Formula Weight: 282.47
Storage Temperature: 2°C to 8°C
Physical Form: Clear liquid
Density: 0.887
Refractive Index: 1.4610
Sensitivity: Air Sensitive
Appearance: Colorless
Melting Point: 13-14°
Boiling Point: 286°/100mm
Beilstein Reference: 1726542
Merck Reference: 14,6828
MDL No.: MFCD00064242
Formula:
C₁₈H₃₄O₂ Boiling Pt: 286 °C/100 mm Melting Pt: 13…14 °C Density: 0.887 Flash Pt: 180 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064242 CAS Number: 112-80-1 EINECS: 204-007-1 Merck Index: 14,6828 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...