2-Chloro-4-methoxyphenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chlor-4-hydroxyanisole

A16652-06 A16652-14
CAAAA16652-06EA 63.2 CAD
CAAAA16652-06 CAAAA16652-14
2-Chloro-4-methoxyphenol 97%
2-Chloro-4-methoxyphenol

Purity: 97% CAS No.: 18113-03-6 Molecular Formula: C7H7ClO2 Formula Weight: 158.59 Flash Point: 85°(185°F) Odor: Phenol-like Beilstein Reference: 1936412 MDL No.: MFCD00070773

Formula: C₇H₇ClO₂
MW: 158.58 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 39…44 °C
Density: 1.153
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070773
CAS Number: 18113-03-6
EINECS: 242-007-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR