2-Fluoro-p-toluidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Fluoro-4-methylaniline, 4-Amino-3-fluorotoluene
A16591-06
A16591-14
CAAAA16591-06EA
170.64
CAD
CAAAA16591-06
CAAAA16591-14
2-Fluoro-p-toluidine 99%
2-Fluoro-p-toluidine
Purity: 99%
CAS No.: 452-80-2
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 79°(174°F)
Refractive Index: 1.5330
Boiling Point: 70-71°/7mm
Beilstein Reference: 2637578
UN No.: 2810
MDL No.: MFCD00040975
Formula:
C₇H₈FN Boiling Pt: 70…71 °C/7 mm Melting Pt: 3 °C Density: 1.108 Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040975 CAS Number: 452-80-2 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...