2-Chloro-4-nitroaniline 98+%

Supplier: Thermo Scientific Chemicals
Warning

A16586-0B A16586-0I A16586-30
CAAAA16586-0BEA 248.06 CAD
CAAAA16586-0B CAAAA16586-0I CAAAA16586-30
2-Chloro-4-nitroaniline 98+%
2-Chloro-4-nitroaniline

Purity: 98+% CAS No.: 121-87-9 Molecular Formula: C6H5ClN2O2 Formula Weight: 172.57 Flash Point: 193°(379°F) Odor: Weak, characteristic Boiling Point: 320° dec. Beilstein Reference: 638657 UN No.: 2237 MDL No.: MFCD00007665

Formula: C₆H₅ClN₂O₂
Boiling Pt: 320 °C (dec.)
Melting Pt: 106…109 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 193 °C (379 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007665
CAS Number: 121-87-9
EINECS: 204-502-2
UN: 2237
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR