(±)-1-Aminoindan 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Aminoindan, 1-Indanamine, 2,3-Dihydro-1H-inden-1-amine, (±)-1-Indanylamine
A16585-03
A16585-06
CAAAAA16585-03EA
67.31
CAD
CAAAAA16585-03
CAAAA16585-06
(±)-1-Aminoindan 99%
(±)-1-Aminoindan
Purity: 99%
CAS No.: 34698-41-4
Molecular Formula: C9H11N
Formula Weight: 133.19
Flash Point: 94°(201°F)
Refractive Index: 1.5620
Melting Point: 2°
Boiling Point: 225°
Beilstein Reference: 3196205
MDL No.: MFCD00003799
Formula:
C₉H₁₁N MW: 133.19 g/mol Boiling Pt: 225 °C Melting Pt: 2 °C Density: 1.038 Flash Pt: 94 °C(201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003799 CAS Number: 34698-41-4 EINECS: 252-158-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...