Propagyl bromide ≥80% in toluene stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabiised with magnesium oxide (MgO)
A16580-AD
A16580-AK
A16580-AU
CAAAA16580-ADEA
154.84
CAD
CAAAA16580-AD
CAAAA16580-AK
CAAAA16580-AU
Propagyl bromide ≥80% in toluene stabilized
Propagyl bromide
CAS No.: 106-96-7
Molecular Formula: HC≡CCH2Br
Formula Weight: 118.97
Storage Temperature: 2°C to 8°C
Flash Point: 3°(37°F)
Refractive Index: 1.4940
Melting Point: -61°
Boiling Point: 88-90°
Beilstein Reference: 605309
UN No.: 2924
MDL No.: MFCD00000241
Formula:
C₃H₃Br Boiling Pt: 88…90 °C Melting Pt: –61 °C Density: 1.335 Flash Pt: 3 °C (37 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000241 CAS Number: 106-96-7 EINECS: 203-447-1 UN: 3286 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...