4-Butylbenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Butylbenzene-1-sulphonyl chloride, 4-Butylbenzene-1-sulfonyl chloride

A16577-06 A16577-14
CAAAA16577-06EA 122.92 CAD
CAAAA16577-06 CAAAA16577-14
4-Butylbenzenesulfonyl chloride 97%
4-Butylbenzenesulphonyl chloride

Purity: 97% CAS No.: 54997-92-1 Molecular Formula: C10H13ClO2S Formula Weight: 232.73 Melting Point: 13-15° Boiling Point: 126-128°/0.6mm UN No.: 3265 MDL No.: MFCD00173908

Formula: C₁₀H₁₃ClO₂S
MW: 232.73 g/mol
Boiling Pt: 126…128 °C/0.6 mm
Melting Pt: 13…15 °C
Storage Temperature: Ambient
MDL Number: MFCD00173908
CAS Number: 54997-92-1
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR