Lithium benzoate 99+%

Supplier: Thermo Scientific Chemicals
Warning

A16571-0B A16571-30
CAAAA16571-0BEA 965.38 CAD
CAAAA16571-0B CAAAA16571-0I CAAAA16571-30
Lithium benzoate 99+%
Lithium benzoate

MDL: MFCD00035540 Beilstein: 3598089 Solubility in water is increased by sodium benzoate


Purity: 99+% CAS No.: 553-54-8 Molecular Formula: C7H5LiO2 Formula Weight: 128.06 Odor: Odorless Melting Point: 300° Beilstein Reference: 3598089 Merck Reference: 14,5523 MDL No.: MFCD00035540

Formula: C₇H₅LiO₂
Melting Pt: >300 °C
Density: 1.397 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00035540
CAS Number: 553-54-8
EINECS: 209-042-6
Merck Index: 14,5523

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR