Sodium oxamate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Oxamic acid sodium salt, Sodium 2-amino-2-oxoacetate

A16532-06 A16532-14
CAAAA16532-06EA 72.05 CAD
CAAAA16532-06 CAAAA16532-14
Sodium oxamate 98%
Sodium oxamate

Purity: 98% CAS No.: 565-73-1 Molecular Formula: H2NC(O)CO2Na Formula Weight: 111.03 Melting Point: 300° Beilstein Reference: 6538153

Formula: C₂H₂NNaO₃
MW: 111.03 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00044553
CAS Number: 565-73-1
EINECS: 209-290-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR