2,5-Difluorobenzylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16530-14
A16530-03
A16530-06
CAAAAA16530-14EA
575.12
CAD
CAAAAA16530-14
CAAAA16530-03
CAAAA16530-06
2,5-Difluorobenzylamine 97%
Purity: 97%
CAS No.: 85118-06-5
Molecular Formula: C7H7F2N
Formula Weight: 143.14
Flash Point: 75°(167°F)
Refractive Index: 1.4950
Boiling Point: 76-78°/15mm
Beilstein Reference: 3539280
UN No.: 2735
MDL No.: MFCD00010143
Formula:
C₇H₇F₂N Boiling Pt: 76…78 °C/15 mm Density: 1.222 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010143 CAS Number: 85118-06-5 EINECS: 285-658-9 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...