α-Hydroxy-o-tolyl-β-D-glucopyranoside 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, D(-)-Salicin, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol, D(−)-Salicin
An analgesic and antipyretic. Substrate for beta-glucosidase
Purity: 99%
CAS No.: 138-52-3
Molecular Formula: C13H18O7
Formula Weight: 286.28
Physical Form: Powder
Appearance: White
Odor: Odorless
Melting Point: 200-203°
Beilstein Reference: 89593
Merck Reference: 14,8324
Optical Rotation: -62.5° (c=3 in water)
MDL No.: MFCD00006590
An analgesic and antipyretic. Substrate for beta-glucosidase
Formula:
C₁₃H₁₈O₇ MW: 286.28 g/mol Melting Pt: 200…203 °C Density: 1.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006590 CAS Number: 138-52-3 EINECS: 205-331-6 Merck Index: 14,8324 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...