Methyl carbamate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Carbamic acid methyl ester

A16523-18 A16523-22 A16523-36
CAAAAA16523-18EA 37.92 CAD
CAAAAA16523-18 CAAAA16523-22 CAAAA16523-36
Methyl carbamate 99%
Methyl carbamate

Purity: 99% CAS No.: 598-55-0 Molecular Formula: H2NCO2CH3 Formula Weight: 75.07 Flash Point: 100°(212°F) Melting Point: 54-56° Boiling Point: 176-178° Beilstein Reference: 635779 Merck Reference: 14,6036 MDL No.: MFCD00007964

Formula: C₂H₅NO₂
MW: 75.07 g/mol
Boiling Pt: 176…178 °C
Melting Pt: 54…56 °C
Density: 1.140
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007964
CAS Number: 598-55-0
EINECS: 209-939-2
Merck Index: 14,6036

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR