3,5-Dinitrobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A16502-0B A16502-30
CAAAA16502-0BEA 325.48 CAD
CAAAA16502-0B CAAAA16502-30
3,5-Dinitrobenzoic acid 98+%
3,5-Dinitrobenzoic acid

Purity: 98+% CAS No.: 99-34-3 Molecular Formula: C7H4N2O6 Formula Weight: 212.12 Melting Point: 204-208° Beilstein Reference: 1914286 Merck Reference: 14,3276 UN No.: 1325 MDL No.: MFCD00007253

Formula: C₇H₄N₂O₆
Melting Pt: 204…208 °C
Density: 1.680
Storage Temperature: Ambient
MDL Number: MFCD00007253
CAS Number: 99-34-3
EINECS: 202-751-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,3276

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR