4-Chlorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16325-22
A16325-36
CAAAA16325-22EA
56.25
CAD
CAAAA16325-22
CAAAA16325-36
4-Chlorobenzoyl chloride 98%
4-Chlorobenzoyl chloride
Purity: 98%
CAS No.: 122-01-0
Molecular Formula: C7H4Cl2O
Formula Weight: 175.01
Flash Point: 105°(221°F)
Refractive Index: 1.5780
Odor: Acrid
Melting Point: 12-14°
Boiling Point: 221-222°
Beilstein Reference: 471606
UN No.: 3265
MDL No.: MFCD00000686
Formula:
C₇H₄Cl₂O Boiling Pt: 221…222 °C Melting Pt: 12…14 °C Density: 1.377 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000686 CAS Number: 122-01-0 EINECS: 204-515-3 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...