3,4-Dichlorophenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

A16298-06 A16298-14
CAAAA16298-06EA 73 CAD
CAAAA16298-06 CAAAA16298-14
3,4-Dichlorophenyl isocyanate 97%
3,4-Dichlorophenyl isocyanate

Purity: 97% CAS No.: 102-36-3 Molecular Formula: C7H3Cl2NO Formula Weight: 188.01 Flash Point: 123°(253°F) Odor: Irritating Boiling Point: 118-120°/18mm Beilstein Reference: 608325 UN No.: 2250 MDL No.: MFCD00002017

Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
Boiling Pt: 118…120 °C/18 mm
Melting Pt: 40…44 °C
Flash Pt: 123 °C (253 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002017
CAS Number: 102-36-3
EINECS: 203-026-2
UN: 2250
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR