Tetramethyl-1,3-cyclobutanedione 99%

Supplier: Thermo Scientific Chemicals

A16297-09 A16297-18 A16297-30
CAAAA16297-09EA 77.26 CAD
CAAAA16297-09 CAAAA16297-18 CAAAA16297-30
Tetramethyl-1,3-cyclobutanedione 99%
Tetramethyl-1,3-cyclobutanedione

Purity: 99% CAS No.: 933-52-8 Molecular Formula: C8H12O2 Formula Weight: 140.18 Melting Point: 112-115° Boiling Point: 120-122°/150mm MDL No.: MFCD00001331

Formula: C₈H₁₂O₂
Boiling Pt: 120…122 °C/150 mm
Melting Pt: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00001331
CAS Number: 933-52-8
EINECS: 213-269-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR