Canada balsam (natural), natural filtered

Supplier: Thermo Scientific Chemicals
Warning

A16289-14 A16289-22 A16289-36
CAAAA16289-14EA 66.99 CAD
CAAAA16289-14 CAAAA16289-22 CAAAA16289-36
Canada balsam (natural), natural filtered
Canada balsam

Mounting medium for microscopy


CAS No.: 8007-47-4 Flash Point: 43°(109°F) Refractive Index: 1.5230 Merck Reference: 14,946 UN No.: 1993 MDL No.: MFCD00132800


Mounting medium for microscopy

Density: 0.99
Flash Pt: 43 °C(109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00132800
CAS Number: 8007-47-4
EINECS: 232-362-2
ADR: 3,III
Merck Index: 14,946

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR