2-Diethylaminoethanol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(Diethylamino)-ethanolN,N,-DiethylethanolamineN,N-Diethylethanolamine

A16274-AU A16274-AU
CAAAA16274-AUEA 334.96 CAD
CAAAA16274-AU AAA16274-AU
2-Diethylaminoethanol 99%
2-Diethylaminoethanol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 161…163 °C
Melting Pt: –70 °C
Density: 0.883
Flash Pt: 52 °C (125 °F)
MDL Number: MFCD00002850
CAS Number: 100-37-8
UN: 2686
ADR: 8,II
Merck Index: 14,3112

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR