D-(+)-Maltose monohydrate 95%
Supplier: Thermo Scientific Chemicals
A16266-22
A16266-36
CAAAA16266-22EA
72.52
CAD
CAAAA16266-22
CAAAA16266-36
D-(+)-Maltose monohydrate 95%
D(+)-Maltose monohydrate
MDL: MFCD00149343
Beilstein: 5784659
Soluble in water. Slightly soluble in alcohol. Insoluble in ether
Purity: 95%
CAS No.: 6363-53-7
Molecular Formula: C12H22O11.H2O
Formula Weight: 360.32 (342.30anhy)
Melting Point: ca 120° dec.
Solubility: Soluble in water. Slightly soluble in alcohol. Insoluble in ether
Beilstein Reference: 5784659
Merck Reference: 14,5714
Optical Rotation: +130° (c=2 in water, 24h)
MDL No.: MFCD00149343
Formula:
C₁₂H₂₂O₁₁·1H₂O Melting Pt: ∼ 120 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149343 CAS Number: 6363-53-7 EINECS: 200-716-5 Merck Index: 14,5714 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...