Hexanal 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16265-22
A16265-36
CAAAA16265-22EA
54.19
CAD
CAAAA16265-22
CAAAA16265-36
Hexanal 98%
Hexanal
Purity: 98%
CAS No.: 66-25-1
Molecular Formula: CH3(CH2)4CHO
Formula Weight: 100.16
Flash Point: 25°(77°F)
Refractive Index: 1.4050
Melting Point: -56°
Boiling Point: 130-131°
Beilstein Reference: 506198
Merck Reference: 14,1760
UN No.: 1207
MDL No.: MFCD00007027
Formula:
C₆H₁₂O Boiling Pt: 130…131 °C Melting Pt: –56 °C Density: 0.834 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007027 CAS Number: 66-25-1 EINECS: 200-624-5 UN: 1207 ADR: 3,III Merck Index: 14,1760 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...