1,10-Phenanthroline hydrochloride monohydrate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16254-06
A16254-14
A16254-22
CAAAA16254-06EA
79.62
CAD
CAAAA16254-06
CAAAA16254-14
CAAAA16254-22
1,10-Phenanthroline hydrochloride monohydrate 99%
1,10-Phenanthroline hydrochloride monohydrate
Purity: 99%
CAS No.: 18851-33-7
Molecular Formula: C12H8N2.HCl.H2O
Formula Weight: 234.68 (216.66anhy)
Odor: Odorless
Melting Point: ca 225° dec.
Beilstein Reference: 4007967
UN No.: 2811
MDL No.: MFCD00150061
Formula:
C₁₂H₉ClN₂·1H₂O Melting Pt: ∼ 225 °C (dec.) Density: 1.385 g/cm³ (-100.15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150061 CAS Number: 18851-33-7 EINECS: 223-325-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...