2,4-Dimethoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A16252-14 A16252-22
CAAAA16252-14EA 104.91 CAD
CAAAA16252-14 CAAAA16252-22
2,4-Dimethoxybenzoic acid 98%
2,4-Dimethoxybenzoic acid

Purity: 98% CAS No.: 91-52-1 Molecular Formula: C9H10O4 Formula Weight: 182.18 Odor: Odorless Melting Point: 107-111° Beilstein Reference: 2210860 MDL No.: MFCD00002434

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 313
Melting Pt: 107…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00002434
CAS Number: 91-52-1
EINECS: 202-074-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR