L(+)-Lysine monohydrochloride 99+%
Supplier: Thermo Scientific Chemicals
A16249-18
A16249-0B
A16249-30
CAAAAA16249-18EA
35.08
CAD
CAAAAA16249-18
CAAAA16249-0B
CAAAA16249-30
L(+)-Lysine monohydrochloride 99+%
L(+)-Lysine monohydrochloride
Purity: 99+%
CAS No.: 657-27-2
Molecular Formula: C6H14N2O2.HCl
Formula Weight: 182.65
Melting Point: ca 260° dec.
Beilstein Reference: 3563889
Merck Reference: 14,5636
Optical Rotation: +20° (c=5 in 5N HCl)
MDL No.: MFCD00064564
Formula:
H₂N(CH₂)₄CH(NH₂)CO₂H·HCl Melting Pt: ∼ 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064564 CAS Number: 657-27-2 EINECS: 211-519-9 Merck Index: 14,5636 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...