Vinyl acetate 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with hydroquinone (HQ) 8-12 ppm
A16247-AE
A16247-0F
A16247-AP
CAAAAA16247-AEEA
48.03
CAD
CAAAAA16247-AE
CAAAA16247-0F
CAAAA16247-AP
Vinyl acetate 99% stabilized
Vinyl acetate
Purity: 99%
CAS No.: 108-05-4
Molecular Formula: C4H6O2
Formula Weight: 86.09
Storage Temperature: 2°C to 8°C
Flash Point: -8°(18°F)
Physical Form: Clear liquid
Density: 0.932
Refractive Index: 1.3960
Appearance: Colorless
Odor: Sweet
Melting Point: -100°
Boiling Point: 72-73°
Beilstein Reference: 1209327
Merck Reference: 14,9992
UN No.: 1301
MDL No.: MFCD00008713
Formula:
C₄H₆O₂ Boiling Pt: 72…73 °C Melting Pt: –100 °C Density: 0.932 Flash Pt: –8 °C (18 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008713 CAS Number: 108-05-4 EINECS: 203-545-4 UN: 1301 ADR: 3,II Merck Index: 14,9992 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...