Magnesium stearate

Supplier: Thermo Scientific Chemicals
Warning

A16244-0B A16244-30
CAAAA16244-0BEA 263.86 CAD
CAAAA16244-0B CAAAA16244-30
Magnesium stearate
Magnesium stearate

CAS No.: 557-04-0 Molecular Formula: [CH3(CH2)16CO2]2Mg Formula Weight: 591.27 Physical Form: Powder Odor: Odorless Melting Point: 148-152° MDL No.: MFCD00036391

Formula: C₃₆H₇₀MgO₄
Melting Pt: 148…152 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036391
CAS Number: 557-04-0
EINECS: 209-150-3
Merck Index: 14,5690

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR