Polyethylene glycol 1,500
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16241-0B
A16241-30
CAAAA16241-0BEA
144.73
CAD
CAAAA16241-0B
CAAAA16241-30
Polyethylene glycol 1,500
Polyethylene glycol 1,500
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Odor: Odorless
pH: 5-7
Melting Point: 44-48°
Merck Reference: 14,7568
MDL No.: MFCD01779609
Formula:
H(OCH₂CH₂)nOH Boiling Pt: ? 250 °C (1013 hPa) Melting Pt: 44…48 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,7568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...