4'-Fluoropropiophenone 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Fluoropropiophenone, 1-(4-Fluorophenyl)propan-1-one

A16219-09 A16219-18
CAAAA16219-09EA 46.77 CAD
CAAAA16219-09 CAAAA16219-18
4'-Fluoropropiophenone 97+%
4'-Fluoropropiophenone

Purity: 97+% CAS No.: 456-03-1 Molecular Formula: C9H9FO Formula Weight: 152.17 Flash Point: 76°(168°F) Refractive Index: 1.5059 Boiling Point: 100-102°/22mm Beilstein Reference: 1210310 MDL No.: MFCD00000356

Formula: C₉H₉FO
MW: 152.17
Boiling Pt: 100…102 °C (22 mmHg)
Density: 1.09
Flash Pt: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000356
CAS Number: 456-03-1
EINECS: 207-255-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR