2-Bromo-4-chlorophenol 98+%

Supplier: Thermo Scientific Chemicals
Danger

A16218-14 A16218-22
CAAAA16218-14EA 61.3 CAD
CAAAA16218-14 CAAAA16218-22
2-Bromo-4-chlorophenol 98+%
2-Bromo-4-chlorophenol

Purity: 98+% CAS No.: 695-96-5 Molecular Formula: C6H4BrClO Formula Weight: 207.46 Flash Point: 110°(230°F) Odor: Phenol-like Boiling Point: 230-232° Beilstein Reference: 2042871 MDL No.: MFCD00002319

Formula: C₆H₄BrClO
Boiling Pt: 230…232 °C
Melting Pt: 31…35 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002319
CAS Number: 695-96-5
EINECS: 211-785-6
UN: 2020
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR