1,4-Diiodobutane 99% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with copper

A16216-14 A16216-22
CAAAA16216-14EA 136.04 CAD
CAAAA16216-14 CAAAA16216-22
1,4-Diiodobutane 99% stabilized
1,4-Diiodobutane

Purity: 99% CAS No.: 628-21-7 Molecular Formula: I(CH2)4I Formula Weight: 309.92 Refractive Index: 1.6212 Melting Point: 5-7° Boiling Point: 108-110°/10mm Beilstein Reference: 1098276 MDL No.: MFCD00001099

Formula: C₄H₈I₂
Boiling Pt: 108…110 °C/10 mm
Melting Pt: 5…7 °C
Density: 2.350
Storage Temperature: Ambient
MDL Number: MFCD00001099
CAS Number: 628-21-7
EINECS: 211-032-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR