2,6-Dichloro-4-fluorophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16193-06
A16193-22
CAAAA16193-06EA
67.31
CAD
CAAAA16193-06
CAAAA16193-14
CAAAA16193-22
2,6-Dichloro-4-fluorophenol 98%
2,6-Dichloro-4-fluorophenol
Purity: 98%
CAS No.: 392-71-2
Molecular Formula: C6H3Cl2FO
Formula Weight: 180.99
Flash Point: 103°(217°F)
Odor: Phenol-like
Boiling Point: 142°/100mm
Beilstein Reference: 2046856
UN No.: 3261
MDL No.: MFCD00010675
Formula:
C₆H₃Cl₂FO Boiling Pt: 142 °C/100 mm Melting Pt: 47…51 °C Flash Pt: 103 °C (217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010675 CAS Number: 392-71-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...