Malachite green oxalate
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16186-14
A16186-22
CAAAA16186-14EA
72.54
CAD
CAAAA16186-14
CAAAA16186-22
Malachite green oxalate
Malachite green oxalate
Biological stain, dye reagent, acid base indicator: pH 0.0 yellow, 2.0 green;11.6 green, 14 colorless
CAS No.: 2437-29-8
Molecular Formula: C52H54N4O12
Formula Weight: 927.02
Physical Form: Crystalline powder
Appearance: Green
Melting Point: 164° dec.
Beilstein Reference: 3644550
Merck Reference: 14,5699
UN No.: 2811
MDL No.: MFCD00011766
Biological stain, dye reagent, acid base indicator: pH 0.0 yellow, 2.0 green;11.6 green, 14 colorless
Formula:
C₅₂H₅₄N₄O₁₂ Melting Pt: 164 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011766 CAS Number: 2437-29-8 EINECS: 219-441-7 UN: 2811 ADR: 6.1,III Merck Index: 14,5699 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...