Benzyl salicylate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Benzyl 2-hydroxybenzoate
A16184-0E
A16184-22
A16184-36
CAAAA16184-0EEA
325.48
CAD
CAAAA16184-0E
CAAAA16184-22
CAAAA16184-36
Benzyl salicylate 99%
Benzyl salicylate
Purity: 99%
CAS No.: 118-58-1
Molecular Formula: C14H12O3
Formula Weight: 228.25
Flash Point: 137°(278°F)
Refractive Index: 1.5804
Melting Point: 18-20°
Boiling Point: 190°/14mm
Beilstein Reference: 2115365
Merck Reference: 14,1144
MDL No.: MFCD00020034
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol Boiling Pt: 208 Melting Pt: 23.9 Density: 1.170 Flash Pt: 137 °C (278 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00020034 CAS Number: 118-58-1 EINECS: 204-262-9 Merck Index: 14,1144 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...