(1S)-(+)-Camphor-10-sulfonic acid, (max. 2% H₂O) 98+% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonic acid, ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, D(+)-Camphor-10-sulfonic acid, (1S)-(+)-2-Oxobornane-10-sulphonic acid, D(+)-Camphor-10-sulphonic acid, (1S)-(+)-7,7-Dimethylbicyclo(2.2.1)-2-heptanon-1-methane sulphonic acid, (1S)-(+)-10-Camphorsulphonic acid, (1S)-(+)-2-Oxobornane-10-sulfonic acid, (1S)-(+)-7,7-Dimethylbicyclo(2.2.1)-2-heptanon-1-methane sulfonic acid, D-Camphorsulphonic acid, (1S)-(+)-10-Camphorsulfonic acid, D-Camphorsulfonic acid
Purity: 98+%
CAS No.: 3144-16-9
Molecular Formula: C10H16O4S
Formula Weight: 232.30
Melting Point: 196-200° dec.
Beilstein Reference: 2809675
Merck Reference: 14,1734
Optical Rotation: +20° (c=2 in water)
UN No.: 3261
MDL No.: MFCD00064157
Formula:
C₁₀H₁₆O₄S MW: 232.3 g/mol Boiling Pt: 375 Melting Pt: 196…200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064157 CAS Number: 3144-16-9 EINECS: 221-554-1 UN: 3261 ADR: 8 III Merck Index: 14,1734 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...