Methyl-3-bromobenzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-bromobenzoate, 3-Bromobenzoic acid methyl ester

A16174-30 A16174-09 A16174-18
CAAAAA16174-30EA 668.34 CAD
CAAAAA16174-30 CAAAA16174-09 CAAAA16174-18
Methyl-3-bromobenzoate 98+%
Methyl 3-bromobenzoate

Purity: 98+% CAS No.: 618-89-3 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Flash Point: 110°(230°F) Refractive Index: 1.5575 Melting Point: 30-33° Boiling Point: 139-140°/35mm Beilstein Reference: 1940810 MDL No.: MFCD00017777

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 139…140 °C/35 mm
Melting Pt: 30…33 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017777
CAS Number: 618-89-3
EINECS: 210-569-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR