1-Iododecane 98% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Decane, 1-iodo-, Decyl iodide
Stabilization:
Stabilised with copper
A16173-14
A16173-18
A16173-30
CAAAAA16173-14EA
72.36
CAD
CAAAAA16173-14
CAAAA16173-18
CAAAA16173-30
1-Iododecane 98% stabilised
1-Iododecane
Purity: 98%
CAS No.: 2050-77-3
Molecular Formula: C10H21I
Formula Weight: 268.18
Flash Point: 96°(204°F)
Refractive Index: 1.4850
Melting Point: -16°
Boiling Point: 264°
Beilstein Reference: 1735228
MDL No.: MFCD00001085
Formula:
C₁₀H₂₁I Boiling Pt: 264 °C Melting Pt: –16 °C Density: 1.257 Flash Pt: 96 °C(204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001085 CAS Number: 2050-77-3 EINECS: 218-104-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...