2-Acetyl-5-methylfuran 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(5-Methylfuran-2-yl)ethanone

A16172-14 A16172-09 A16172-18
CAAAAA16172-14EA 87.69 CAD
CAAAAA16172-14 CAAAA16172-09 CAAAA16172-18
2-Acetyl-5-methylfuran 98+%
2-Acetyl-5-methylfuran

Purity: 98+% CAS No.: 1193-79-9 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 80°(176°F) Refractive Index: 1.5130 Boiling Point: 100-101°/25mm Beilstein Reference: 110853 MDL No.: MFCD00003243

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 100…101 °C (25 mmHg)
Density: 1.068
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003243
CAS Number: 1193-79-9
EINECS: 214-779-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR