L(+)-Rhamnose monohydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal hydrate, 6-Deoxy-L(+)-mannose monohydrate

A16166-06 A16166-14 A16166-22
CAAAA16166-06EA 76.8 CAD
CAAAA16166-06 CAAAA16166-14 CAAAA16166-22
L(+)-Rhamnose monohydrate 99%
L(+)-Rhamnose monohydrate

Purity: 99% CAS No.: 10030-85-0 Molecular Formula: C6H12O5.H2O Formula Weight: 182.19 (164.17anhy) Melting Point: 89-94° Beilstein Reference: 5988592 Merck Reference: 14,8172 Optical Rotation: +8° (c=10 in water, 20h) MDL No.: MFCD00149363

Formula: C₆H₁₂O₅·1H₂O
MW: 182.17 g/mol
Melting Pt: 89…94 °C
Density: 1.47
Storage Temperature: Ambient
MDL Number: MFCD00149363
CAS Number: 10030-85-0
EINECS: 222-793-4
Merck Index: 14,8172

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR