Polyethylene glycol 4,000

Supplier: Thermo Scientific Chemicals

Synonyms: PEG 4,000, PEG 4000

A16151-0B A16151-0I A16151-30
CAAAA16151-0BEA 189.25 CAD
CAAAA16151-0B CAAAA16151-0I CAAAA16151-30
Polyethylene glycol 4,000
Polyethylene glycol 4,000

CAS No.: 25322-68-3 Molecular Formula: H(OCH2CH2)nOH Odor: Odorless pH: 5-7 Melting Point: 58-62° Merck Reference: 14,7568 MDL No.: MFCD01779612

Formula: H(OCH₂CH₂)nOH
Boiling Pt: ? 250 °C (1013 hPa)
Melting Pt: 58…62 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD01779612
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,7568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR