Propionaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Propanal
A16146-AP
A16146-0J
A16146-AU
CAAAAA16146-APEA
63.52
CAD
CAAAAA16146-AP
CAAAA16146-0J
CAAAA16146-AU
Propionaldehyde 97%
Propionaldehyde
Purity: 97%
CAS No.: 123-38-6
Molecular Formula: CH3CH2CHO
Formula Weight: 58.08
Storage Temperature: 2°C to 8°C
Flash Point: -40°(-40°F)
Refractive Index: 1.3620
Odor: Pungent
Melting Point: -81°
Boiling Point: 47-49°
Beilstein Reference: 506010
Merck Reference: 14,7823
UN No.: 1275
MDL No.: MFCD00007020
Formula:
C₃H₆O MW: 58.08 g/mol Boiling Pt: 47…49 °C Melting Pt: –81 °C Density: 0.81 Flash Pt: –40 °C (–40 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007020 CAS Number: 123-38-6 EINECS: 204-623-0 UN: 1275 ADR: 3,II Merck Index: 14,7823 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...