Methyl-2-fluorobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

A16133-06 A16133-14
CAAAA16133-06EA 22.59 CAD
CAAAA16133-06 CAAAA16133-14
Methyl-2-fluorobenzoate 98%
Methyl 2-fluorobenzoate

Purity: 98% CAS No.: 394-35-4 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Flash Point: 93°(199°F) Refractive Index: 1.5030 Odor: Aromatic Boiling Point: 109-110°/35mm Beilstein Reference: 1862493 MDL No.: MFCD00017913

Formula: C₈H₇FO₂
Boiling Pt: 109…110 °C/35 mm
Density: 1.210
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017913
CAS Number: 394-35-4
EINECS: 206-894-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR