L(+)-Lysine dihydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Lys-OH·2HCl, H-Lys-OH.2HCl, (S)-(+)-Lysine dihydrochloride, (S)-(+)-2,6-Diaminohexanoic acid dihydrochloride, (S)-(+)-2,6-Diaminocaproic acid dihydrochloride

A16127-14 A16127-22
CAAAA16127-14EA 49.3 CAD
CAAAA16127-14 CAAAA16127-22
L(+)-Lysine dihydrochloride 99%
L(+)-Lysine dihydrochloride

Purity: 99% CAS No.: 657-26-1 Molecular Formula: C6H14N2O2.2HCl Formula Weight: 219.11 Melting Point: 202-206° Beilstein Reference: 4539715 Merck Reference: 14,5636 MDL No.: MFCD00066389

Formula: C₆H₁₆Cl₂N₂O₂
MW: 219.11 g/mol
Melting Pt: 200.2
Storage Temperature: Ambient
MDL Number: MFCD00066389
CAS Number: 657-26-1
EINECS: 211-518-3
Merck Index: 14,5636

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR