1-Naphtholbenzeine
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
alpha-Naphtholbenzein, p-Naphtholbenzein, α-Naphtholbenzein, p-Naphtholbenzeine, 4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one, 1-Naphtholbenzein, Bis(4-hydroxy-1-naphthyl)phenylmethanol, 4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanol, α-Naphtholbenzeine
A16125-14
A16125-22
CAAAA16125-14EA
226.73
CAD
CAAAA16125-14
CAAAA16125-22
1-Naphtholbenzeine
1-Naphtholbenzeine
CAS No.: 145-50-6
Molecular Formula: C27H18O2
Formula Weight: 374.44
Melting Point: ca. 225° dec.
Beilstein Reference: 3471575
MDL No.: MFCD00078492
Formula:
C₂₇H₁₈O₂ MW: 374.44 g/mol Melting Pt: ∼ 225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00078492 CAS Number: 145-50-6 EINECS: 205-656-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...