5-Bromo-2-fluorobenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16113-06
A16113-14
CAAAA16113-06EA
45.66
CAD
CAAAA16113-06
CAAAA16113-14
5-Bromo-2-fluorobenzonitrile 98%
5-Bromo-2-fluorobenzonitrile
Purity: 98%
CAS No.: 179897-89-3
Molecular Formula: C7H3BrFN
Formula Weight: 200.01
Melting Point: 77-81°
Beilstein Reference: 7701063
UN No.: 3439
MDL No.: MFCD00143424
Formula:
C₇H₃BrFN Boiling Pt: 255 °C (1013 hPa) Melting Pt: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143424 CAS Number: 179897-89-3 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...