4-Bromo-2-fluorobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16112-06
A16112-14
CAAAAA16112-22EA
2208.78
CAD
CAAAAA16112-22
CAAAA16112-06
CAAAA16112-14
4-Bromo-2-fluorobenzaldehyde 97%
4-Bromo-2-fluorobenzaldehyde
Purity: 97%
CAS No.: 57848-46-1
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Melting Point: 59-63°
Boiling Point: 42°/19mm
MDL No.: MFCD00143261
Formula:
C₇H₄BrFO MW: 203.01 g/mol Boiling Pt: 42 °C/19 mm Melting Pt: 59…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143261 CAS Number: 57848-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...