3-Fluorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16107-14
A16107-22
A16107-30
CAAAAA16107-14EA
34.29
CAD
CAAAAA16107-14
CAAAAA16107-22
CAAAA16107-30
3-Fluorotoluene 98%
3-Fluorotoluene
Purity: 98%
CAS No.: 352-70-5
Molecular Formula: C7H7F
Formula Weight: 110.13
Flash Point: 9°(48°F)
Refractive Index: 1.4690
Odor: Aromatic
Melting Point: -87°
Boiling Point: 114-116°
Beilstein Reference: 1903631
Merck Reference: 14,4180
UN No.: 2388
MDL No.: MFCD00000339
Formula:
C₇H₇F Boiling Pt: 114…116 °C Melting Pt: –87 °C Density: 0.993 Flash Pt: 9 °C (48 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000339 CAS Number: 352-70-5 EINECS: 206-524-8 UN: 2388 ADR: 3,II Merck Index: 14,4180 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...