β-(N-Morpholino)ethanesufonic acid (MES) monohydrate 98%
Supplier: Thermo Scientific Chemicals
A16104-14
A16104-22
A16104-36
CAAAA16104-14EA
75.21
CAD
CAAAA16104-14
CAAAA16104-22
CAAAA16104-36
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate 98%
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Good's buffer
Purity: 98%
CAS No.: 145224-94-8
Molecular Formula: C6H13NO4S.H2O
Formula Weight: 213.26 (195.24anhy)
Physical Form: Crystalline powder
Appearance: White
Melting Point: ca 308° dec.
Beilstein Reference: 141319
Merck Reference: 14,5902
MDL No.: MFCD00149409
Good's buffer
Formula:
C₆H₁₃NO₄S·1H₂O Melting Pt: ∼ 308 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149409 CAS Number: 145224-94-8 EINECS: 224-632-3 Merck Index: 14,5902 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...