2-Fluorobenzoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Fluorobenzoyl chloride

A16096-22 A16096-36
CAAAA16096-22EA 173.8 CAD
CAAAA16096-22 CAAAA16096-36
2-Fluorobenzoyl chloride 97%
2-Fluorobenzoyl chloride

Purity: 97% CAS No.: 393-52-2 Molecular Formula: C7H4ClFO Formula Weight: 158.56 Flash Point: 82°(179°F) Refractive Index: 1.5365 Odor: Pungent Melting Point: 4-6° Boiling Point: 207° Beilstein Reference: 636864 UN No.: 3265 MDL No.: MFCD00000656

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 206
Melting Pt: 4…6 °C
Density: 1.331
Flash Pt: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000656
CAS Number: 393-52-2
EINECS: 206-887-2
UN: 1760
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR